(4-Acetamidophenyl)boronic acid pinacol ester ≥98.0%

Supplier: TCI America

Synonyms: N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acetanilide

T1950-1G
TCT1950-1GEA 77 USD
TCT1950-1G
(4-Acetamidophenyl)boronic acid pinacol ester ≥98.0%
(4-Acetamidophenyl)boronic acid pinacol ester

CAS Number: 214360-60-8
MDL Number: MFCD02093722
Molecular Formula: C14H20BNO3
Molecular Weight: 261.13
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 166


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2-(4-Acetamidophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

https://us.vwr.com/stibo/web/std.lang.all/59/02/28995902.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR