6-O-(Triisopropylsilyl)-D-galactal ≥97.0%

Synonyms: 1,5-Anhydro-2-deoxy-6-O-(triisopropylsilyl)-D-lyxo-hex-1-enitol

T1935-200MG
TCT1935-200MGEA 233.58 USD
TCT1935-200MG
6-O-(Triisopropylsilyl)-D-galactal ≥97.0%
6-O-(Triisopropylsilyl)-D-galactal

CAS Number: 166021-01-8
Molecular Formula: C15H30O4Si
Molecular Weight: 302.49
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 110


Product Categories: Bioscience, Glycals, Galactose
Other Product Names / Synonyms: 1,5-Anhydro-2-deoxy-6-O-(triisopropylsilyl)-D-lyxo-hex-1-enitol

Formula: C₁₅H₃₀O₄Si
MW: 302.49 g/mol
Flash Pt: 110 °C
CAS Number: 166021-01-8
https://us.vwr.com/stibo/web/std.lang.all/58/89/28995889.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR