1,3,5-Tri(9H-carbazol-9-yl)benzene ≥98.0% (by HPLC), purified by sublimation

Supplier: TCI America

T1934-5G T1934-1G
TCT1934-5GEA 897 USD
TCT1934-5G TCT1934-1G
1,3,5-Tri(9H-carbazol-9-yl)benzene ≥98.0% (by HPLC), purified by sublimation
1,3,5-Tri(9H-carbazol-9-yl)benzene

CAS Number: 148044-07-9
MDL Number: MFCD03844810
Molecular Formula: C42H27N3
Molecular Weight: 573.70
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 330

Formula: C₄₂H₂₇N₃
Melting Pt: 330 °C
MDL Number: MFCD03844810
CAS Number: 148044-07-9
https://us.vwr.com/stibo/web/std.lang.all/58/88/28995888.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR