1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,2',2'',2'''-(1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetrayl)tetraacetic acidDOTA1,4,7,10-Tetraazacyclododecane-N,N',N'',N'''-tetraacetic acid

T1875-1G T1875-200MG
TCT1875-200MGEA 159.31 USD
TCT1875-200MG TCT1875-1G
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid ≥97.0% (by titrimetric analysis)
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid

CAS Number: 60239-18-1
MDL Number: MFCD00068657
Molecular Formula: C16H28N4O8
Molecular Weight: 404.42
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 267


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Macrocycles for Host-Guest Chemistry, Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: DOTA

Formula: C₁₆H₂₈N₄O₈
MW: 404.42 g/mol
Melting Pt: 267 °C
MDL Number: MFCD00068657
CAS Number: 60239-18-1
https://us.vwr.com/stibo/web/std.lang.all/58/40/28995840.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR