(m-Terphenyl-5'-yl)trimethylsilane ≥98.0%

Supplier: TCI America

Synonyms: 3,5-Diphenyl-1-trimethylsilylbenzeneTrimethyl(m-terphenyl-5'-yl)silane

T1871-1G
TCT1871-1GEA 182.62 USD
TCT1871-1G
(m-Terphenyl-5'-yl)trimethylsilane ≥98.0%
(m-Terphenyl-5'-yl)trimethylsilane

CAS Number: 128388-53-4
MDL Number: MFCD03844807
Molecular Formula: C21H22Si
Molecular Weight: 302.49
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 81


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 3,5-Diphenyl-1-trimethylsilylbenzene, Trimethyl(m-terphenyl-5'-yl)silane

Formula: C₂₁H₂₂Si
MW: 302.49 g/mol
Melting Pt: 81 °C
MDL Number: MFCD03844807
CAS Number: 128388-53-4
https://us.vwr.com/stibo/web/std.lang.all/58/37/28995837.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR