Methyl-3,4,5-tris(benzyloxy)benzoate ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
3,4,5-Tribenzyloxybenzoic Acid Methyl Ester
, 3,4,5-Tris(benzyloxy)benzoic Acid Methyl Ester
, Methyl 3,4,5-tris(benzyloxy)benzoate
, Methyl 3,4,5-Tribenzyloxybenzoate
CAS Number: 70424-94-1
MDL Number: MFCD03425302
Molecular Formula: C29H26O5
Molecular Weight: 454.52
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 102
Other Product Names / Synonyms: Methyl 3,4,5-Tribenzyloxybenzoate, 3,4,5-Tribenzyloxybenzoic Acid Methyl Ester, 3,4,5-Tris(benzyloxy)benzoic Acid Methyl Ester
Formula:
C₂₉H₂₆O₅ MW: 454.52 g/mol Melting Pt: 102 °C |
MDL Number:
MFCD03425302 CAS Number: 70424-94-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...