Triphenylbismuth diacetate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1836-5G
TCT1836-5GEA 255.82 USD
TCT1836-5G
Triphenylbismuth diacetate ≥98.0% (by titrimetric analysis)
Triphenylbismuth diacetate

CAS Number: 7239-60-3
MDL Number: MFCD00092697
Molecular Formula: C22H21BiO4
Molecular Weight: 558.39
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: Very Pale Yellow


Product Categories: Synthetic Organic Chemistry, Hypervalent Bismuth Compounds
Other Product Names / Synonyms: Bis(acetato-O)triphenylbismuth

Formula: C₂₂H₂₁BiO₄
MDL Number: MFCD00092697
CAS Number: 7239-60-3
https://us.vwr.com/stibo/web/std.lang.all/58/09/28995809.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR