CLA ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
CLA-phenyl
[Chemiluminescence Reagent]
CAS Number: 19953-58-3
MDL Number: MFCD00130107
Molecular Formula: C13H11N3O
Molecular Weight: 225.25
Purity/Analysis Method:
Form: Crystal
Storage Temperature: 0-10°C
Product Categories: Analytical Science, Cypridina Luciferin Analogs (Chemiluminescence Reagents)
Other Product Names / Synonyms: CLA-phenyl, 2-Methyl-6-phenyl-3,7-dihydroimidazo[1,2-a]pyrazin-3-one
Formula:
C₁₃H₁₁N₃O MW: 225.25 g/mol |
MDL Number:
MFCD00130107 CAS Number: 19953-58-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...