7-Tetradecanol ≥96.0%

Supplier: TCI America

T1759-1G
TCT1759-001GEA 65 USD
TCT1759-001G
7-Tetradecanol ≥96.0%
7-Tetradecanol

CAS Number: 3981-79-1
MDL Number: MFCD00049210
Molecular Formula: C14H30O
Molecular Weight: 214.39
Purity/Analysis Method: 96.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 156
Melting point (°C): 42
Flash Point (°C): 66

https://us.vwr.com/stibo/web/std.lang.all/57/36/28995736.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR