N-(1,1,3,3-Tetramethylbutyl)formamide ≥98.0%

Supplier: TCI America

Synonyms: N-Formyl-1,1,3,3-tetramethylbutylaminetert-OctylformamideN-Formyl-tert-octylamine

T1758-5G
TCT1758-005GEA 115.66 USD
TCT1758-005G
N-(1,1,3,3-Tetramethylbutyl)formamide ≥98.0%
N-(1,1,3,3-Tetramethylbutyl)formamide

CAS Number: 10151-02-7
MDL Number: MFCD00039569
Molecular Formula: C9H19NO
Molecular Weight: 157.26
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 110
Specific Gravity (20/20): 0.90


Other Product Names / Synonyms: N-Formyl-tert-octylamine, N-Formyl-1,1,3,3-tetramethylbutylamine, tert-Octylformamide

Formula: C₉H₁₉NO
MW: 157.26 g/mol
Boiling Pt: 110 °C
Density: 0.90 (20 °C)
MDL Number: MFCD00039569
CAS Number: 10151-02-7
https://us.vwr.com/stibo/web/std.lang.all/57/35/28995735.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR