(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol ≥95.0% (by titrimetric analysis)

Synonyms: (S)-3-Hydroxymethyl-1,2,3,4-tetrahydroisoquinoline

T1755-1G
TCT1755-001GEA 110.21 USD
TCT1755-001G
(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol ≥95.0% (by titrimetric analysis)
(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol

CAS Number: 18881-17-9
MDL Number: MFCD01631316
Molecular Formula: C10H13NO
Molecular Weight: 163.22
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Color: White
Melting point (°C): 114
Specific rotation [a]20/D: -97 deg (C=0.2, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-3-Hydroxymethyl-1,2,3,4-tetrahydroisoquinoline

Formula: C₁₀H₁₃NO
MW: 163.22 g/mol
Melting Pt: 114 °C
MDL Number: MFCD01631316
CAS Number: 18881-17-9
https://us.vwr.com/stibo/web/std.lang.all/57/33/28995733.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR