Tetrakis(dimethylsilyl)silane ≥95.0%
Supplier: TCI America
T1753-1G
TCT1753-001GEA
717
USD
TCT1753-001G
Tetrakis(dimethylsilyl)silane ≥95.0%
Tetrakis(dimethylsilyl)silane
CAS Number: 2003-85-2
MDL Number: MFCD01631315
Molecular Formula: C8H28Si5
Molecular Weight: 264.74
Purity/Analysis Method:
Form: Crystal
Color: White
Freezing point (°C): 21
Storage Temperature: 0-10°C
Formula:
C₈H₂₈Si₅ |
MDL Number:
MFCD01631315 CAS Number: 2003-85-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...