5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobiindane ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobisindane

T1737-25G
TCT1737-025GEA 61.51 USD
TCT1737-025G
5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobiindane ≥96.0% (by HPLC, titration analysis)
5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobiindane

CAS Number: 77-08-7
MDL Number: MFCD00021235
Molecular Formula: C21H24O4
Molecular Weight: 340.42
Purity/Analysis Method: 96.0% (HPLC,T)
Form: Crystal
Color: Slightly Pale Yellow


Other Product Names / Synonyms: 5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobisindane

Formula: C₂₁H₂₄O₄
MW: 340.42 g/mol
MDL Number: MFCD00021235
CAS Number: 77-08-7
https://us.vwr.com/stibo/web/std.lang.all/57/18/28995718.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR