1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-ɑ-D-galactopyranose
Supplier: TCI America
T1731-100MG
TCT1731-100MGEA
236
USD
TCT1731-100MG
1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-ɑ-D-galactopyranose
1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-alpha-D-galactopyranose
CAS Number: 67817-30-5
MDL Number: MFCD01076182
Molecular Formula: C14H19N3O9
Molecular Weight: 373.32
Form: Crystal
Color: White
Melting point (°C): 121
Specific rotation [a]20/D: 100 deg (C=1, CHCl3)
Storage Temperature: 0-10°C
Formula:
C₁₄H₁₉N₃O₉ MW: 373.32 g/mol Melting Pt: 121 °C |
MDL Number:
MFCD01076182 CAS Number: 67817-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...