5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: 5,10,15,20-Tetrakis(perfluorophenyl)porphyrin5,10,15,20-Tetrakis(pentafluorophenyl)-21H,23H-porphine

T1730-100MG
TCT1730-100MGEA 165.4 USD
TCT1730-100MG
5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin ≥95.0% (by HPLC)
5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin

CAS Number: 25440-14-6
MDL Number: MFCD00010032
Molecular Formula: C44H10F20N4
Molecular Weight: 974.56
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal
Color: Purple


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Bioscience, Porphyrins
Other Product Names / Synonyms: 5,10,15,20-Tetrakis(perfluorophenyl)porphyrin, 5,10,15,20-Tetrakis(pentafluorophenyl)-21H,23H-porphine

Formula: C₄₄H₁₀F₂₀N₄
MW: 974.56 g/mol
MDL Number: MFCD00010032
CAS Number: 25440-14-6
https://us.vwr.com/stibo/web/std.lang.all/57/13/28995713.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR