2,3,4,4'-Tetrahydroxydiphenylmethane ≥95.0%
Supplier: TCI America
T1707-5G
T1707-25G
TCT1707-005GEA
195
USD
TCT1707-005G
TCT1707-25G
2,3,4,4'-Tetrahydroxydiphenylmethane ≥95.0%
2,3,4,4'-Tetrahydroxydiphenylmethane
CAS Number: 174462-43-2
MDL Number: MFCD01321194
Molecular Formula: C13H12O4
Molecular Weight: 232.24
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 151
Formula:
C₁₃H₁₂O₄ Melting Pt: 151 °C |
MDL Number:
MFCD01321194 CAS Number: 174462-43-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...