2-Trimethylsilylthiophene ≥98.0%

Supplier: TCI America

Synonyms: Trimethyl(thiophen-2-yl)silane2-Thienyltrimethylsilane

T1706-5G
TCT1706-005GEA 65.94 USD
TCT1706-005G
2-Trimethylsilylthiophene ≥98.0%
2-Trimethylsilylthiophene

CAS Number: 18245-28-8
MDL Number: MFCD00005415
Molecular Formula: C7H12SSi
Molecular Weight: 156.32
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 47
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: 2-Thienyltrimethylsilane

Formula: C₇H₁₂SSi
MW: 156.32 g/mol
Density: 0.95 (20 °C)
Flash Pt: 47 °C
MDL Number: MFCD00005415
CAS Number: 18245-28-8
https://us.vwr.com/stibo/web/std.lang.all/56/91/28995691.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR