TCIAT1664

T1664-5ML
TCT1664-005MLEA 77.93 USD
TCT1664-005ML
TCIAT1664
CAS Number: 26767-00-0
MDL Number: MFCD00008457
Molecular Formula: C8H16O3Si
Molecular Weight: 188.30
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 64
Specific Gravity (20/20): 0.97
Storage Temperature: 0-10°C

Other Product Names / Synonyms: Methyl 3-(Trimethylsilyloxy)-2-butenoate, 3-(Trimethylsilyloxy)crotonic Acid Methyl Ester, 3-(Trimethylsilyloxy)-2-butenoic Acid Methyl Ester
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR