4,4'-Bis(dimethylamino)thiobenzophenone ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Thiomichler's ketone
, N,N,N',N'-Tetramethyl-4,4'-diaminothiobenzophenone
, 4,4'-Bis(dimethylamino)thiobenzophenone
[Sensitive spectrophotometric reagent for Au, Ag, Hg and Pd, use for the determination of residual chlorine]
CAS Number: 1226-46-6
MDL Number: MFCD00040477
Molecular Formula: C17H20N2S
Molecular Weight: 284.42
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 204
Other Product Names / Synonyms: 4,4'-Bis(dimethylamino)thiobenzophenone, N,N,N',N'-Tetramethyl-4,4'-diaminothiobenzophenone
Formula:
C₁₇H₂₀N₂S MW: 284.43 g/mol Melting Pt: 204 °C |
MDL Number:
MFCD00040477 CAS Number: 1226-46-6 EINECS: 214-962-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...