Tetraoctylphosphonium bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tetra-n-octylphosphonium bromide

T1650-25G
TCT1650-025GEA 146 USD
TCT1650-025G
Tetraoctylphosphonium bromide ≥98.0% (by titrimetric analysis)
Tetraoctylphosphonium bromide

CAS Number: 23906-97-0
MDL Number: MFCD00210101
Molecular Formula: C32H68BrP
Molecular Weight: 563.77
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 42
Flash Point (°C): 121
Storage Temperature: 0-10°C

Formula: C₃₂H₆₈BrP
MW: 563.77 g/mol
Melting Pt: 42 °C
Flash Pt: 121 °C
MDL Number: MFCD00210101
CAS Number: 23906-97-0
https://us.vwr.com/stibo/web/std.lang.all/56/41/28995641.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR