2,3,4,4'-Tetrahydroxybenzophenone ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (4-Hydroxyphenyl)(2,3,4-trihydroxyphenyl)methanone

T1594-25G
TCT1594-025GEA 62 USD
TCT1594-025G
2,3,4,4'-Tetrahydroxybenzophenone ≥98.0% (by titrimetric analysis)
2,3,4,4'-Tetrahydroxybenzophenone

CAS Number: 31127-54-5
MDL Number: MFCD00083097
Molecular Formula: C13H10O5
Molecular Weight: 246.22
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 221

Formula: C₁₃H₁₀O₅
MW: 246.2155 g/mol
Melting Pt: 221 °C
MDL Number: MFCD00083097
CAS Number: 31127-54-5
https://us.vwr.com/stibo/web/std.lang.all/56/01/28995601.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR