5-(Trimethylsilyl)thiazole ≥98.0%

Supplier: TCI America

T1523-1ML
TCT1523-001MLEA 81 USD
TCT1523-001ML
5-(Trimethylsilyl)thiazole ≥98.0%
5-(Trimethylsilyl)thiazole

CAS Number: 79265-36-4
MDL Number: MFCD00191506
Molecular Formula: C6H11NSSi
Molecular Weight: 157.31
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 65
Specific Gravity (20/20): 1.00

Formula: C₆H₁₁NSSi
MW: 157.31 g/mol
Boiling Pt: 65 °C
Density: 1.00 (20 °C)
MDL Number: MFCD00191506
CAS Number: 79265-36-4

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR