5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: 5,10,15,20-Tetrakis(4-hydroxyphenyl)-21H,23H-porphine

T1497-100MG
TCT1497-100MGEA 77.14 USD
TCT1497-100MG
5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin ≥95.0% (by HPLC)
5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin

CAS Number: 51094-17-8
MDL Number: MFCD00191501
Molecular Formula: C44H30N4O4
Molecular Weight: 678.75
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal
Color: Purple


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Bioscience, Porphyrins
Other Product Names / Synonyms: 5,10,15,20-Tetrakis(4-hydroxyphenyl)-21H,23H-porphine

Formula: C₄₄H₃₀N₄O₄
MW: 678.75 g/mol
MDL Number: MFCD00191501
CAS Number: 51094-17-8
https://us.vwr.com/stibo/web/std.lang.all/55/14/28995514.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR