Ethyl violet ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Basic Violet 4
A5101-1G
TCA5101-001GEA
72.07
USD
TCA5101-001G
Ethyl violet ≥98.0% (by titrimetric analysis)
Ethyl violet
[Sensitive spectrophotometric reagent for anionic surfactants]
CAS Number: 2390-59-2
MDL Number: MFCD00011765
Molecular Formula: C31H42ClN3
Molecular Weight: 560.29
Purity/Analysis Method:
Form: Crystal
Lambda max.: 596 nm (H2O)
Product Categories: Bioscience, Stains & Dyes, Analytical Science, Ion Association Reagents
Other Product Names / Synonyms: Basic Violet 4
Formula:
C₃₁H₄₂ClN₃ MW: 560.29 g/mol |
MDL Number:
MFCD00011765 CAS Number: 2390-59-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...