(S)-(-)-1,1,2-Triphenyl-1,2-ethanediol
Supplier: TCI America
Synonyms:
(S)-(-)-Triphenylethylene Glycol
T1482-1G
TCT1482-001GEA
66
USD
TCT1482-001G
(S)-(-)-1,1,2-Triphenyl-1,2-ethanediol
(S)-(-)-1,1,2-Triphenyl-1,2-ethanediol
CAS Number: 108998-83-0
MDL Number: MFCD00075492
Molecular Formula: C20H18O2
Molecular Weight: 290.36
Form: Crystal
Color: White
Melting point (°C): 128
Other Product Names / Synonyms: (S)-(-)-Triphenylethylene Glycol
Formula:
C₂₀H₁₈O₂ MW: 290.36 g/mol Melting Pt: 128 °C |
MDL Number:
MFCD00075492 CAS Number: 108998-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...