3,3,3',3'-Tetramethyl-5,5',6,6'-tetrapropoxy-1,1'-spirobiindane ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 3,3,3',3'-Tetramethyl-5,5',6,6'-tetrapropoxy-1,1'-spirobisindane

T1477-25G
TCT1477-025GEA 115.66 USD
TCT1477-025G
3,3,3',3'-Tetramethyl-5,5',6,6'-tetrapropoxy-1,1'-spirobiindane ≥98.0% (by HPLC)
3,3,3',3'-Tetramethyl-5,5',6,6'-tetrapropoxy-1,1'-spirobiindane

CAS Number: 89929-65-7
MDL Number: MFCD00191503
Molecular Formula: C33H48O4
Molecular Weight: 508.74
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 82


Other Product Names / Synonyms: 3,3,3',3'-Tetramethyl-5,5',6,6'-tetrapropoxy-1,1'-spirobisindane

Formula: C₃₃H₄₈O₄
MW: 508.74 g/mol
Melting Pt: 82 °C
MDL Number: MFCD00191503
CAS Number: 89929-65-7
https://us.vwr.com/stibo/web/std.lang.all/54/97/28995497.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR