1,4,5,6-Tetrahydro-5,6-dioxo-2,3-pyrazinedicarbonitrile ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2,3-Dicyano-1,4,5,6-tetrahydropyrazine-5,6-dione
T1472-10G
TCT1472-010GEA
135
USD
TCT1472-010G
1,4,5,6-Tetrahydro-5,6-dioxo-2,3-pyrazinedicarbonitrile ≥98.0% (by HPLC, titration analysis)
1,4,5,6-Tetrahydro-5,6-dioxo-2,3-pyrazinedicarbonitrile
CAS Number: 36023-64-0
MDL Number: MFCD00191690
Molecular Formula: C6H2N4O2
Molecular Weight: 162.11
Purity/Analysis Method:
Form: Crystal
Color: Very Pale Yellow
Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 2,3-Dicyano-1,4,5,6-tetrahydropyrazine-5,6-dione
Formula:
C₆H₂N₄O₂ MW: 162.11 g/mol |
MDL Number:
MFCD00191690 CAS Number: 36023-64-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...