2,4,6-Triphenylpyrylium hydrogensulfate ≥97.0% (by HPLC)

Supplier: TCI America

T1445-1G
TCT1445-001GEA 295 USD
TCT1445-001G
2,4,6-Triphenylpyrylium hydrogensulfate ≥97.0% (by HPLC)
2,4,6-Triphenylpyrylium hydrogensulfate

CAS Number: 51071-75-1
MDL Number: MFCD00012002
Molecular Formula: C23H18O5S
Molecular Weight: 406.45
Purity/Analysis Method: 97.0% (HPLC)
Form: Crystal
Color: Yellow
Melting point (°C): 271

Formula: C₂₃H₁₈O₅S
MW: 406.45 g/mol
Melting Pt: 271 °C
MDL Number: MFCD00012002
CAS Number: 51071-75-1
https://us.vwr.com/stibo/web/std.lang.all/54/64/28995464.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR