N-(p-Toluenesulfonyl)-L-phenylalanylchloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tosyl-L-phenylalanyl Chloride

T1444-5G
TCT1444-005GEA 121.77 USD
TCT1444-005G
N-(p-Toluenesulfonyl)-L-phenylalanylchloride ≥97.0% (by titrimetric analysis)
N-(p-Toluenesulfonyl)-L-phenylalanylchloride

[Optical Resolving Reagent for Alcohols]
CAS Number: 29739-88-6
MDL Number: MFCD00191583
Molecular Formula: C16H16ClNO3S
Molecular Weight: 337.82
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 132
Specific rotation [a]20/D: 10 deg (C=3, CHCl3)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents
Other Product Names / Synonyms: Tosyl-L-phenylalanyl Chloride

Formula: C₁₆H₁₆ClNO₃S
MW: 337.82 g/mol
Melting Pt: 132 °C
MDL Number: MFCD00191583
CAS Number: 29739-88-6
https://us.vwr.com/stibo/web/std.lang.all/54/63/28995463.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR