D-Talitol ≥98.0% (by HPLC)

Supplier: TCI America

T1398-100MG
TCT1398-100MGEA 225.27 USD
TCT1398-100MG
D-Talitol ≥98.0% (by HPLC)
D-Talitol

CAS Number: 643-03-8
MDL Number: MFCD00191491
Molecular Formula: C6H14O6
Molecular Weight: 182.17
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 88
Specific rotation [a]20/D: 2.8 deg (C=1, H2O)

Formula: C₆H₁₄O₆
Melting Pt: 88 °C
MDL Number: MFCD00191491
CAS Number: 643-03-8
https://us.vwr.com/stibo/web/std.lang.all/54/21/28995421.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR