1-(p-Toluenesulfonyl)pyrrole ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-Tosylpyrrole1-Tosylpyrrole

T1386-5G
TCT1386-005GEA 33.5 USD
TCT1386-005G
1-(p-Toluenesulfonyl)pyrrole ≥98.0% (by titrimetric analysis)
1-(p-Toluenesulfonyl)pyrrole

CAS Number: 17639-64-4
MDL Number: MFCD00145014
Molecular Formula: C11H11NO2S
Molecular Weight: 221.27
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 103


Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 1-Tosylpyrrole

Formula: C₁₁H₁₁NO₂S
MW: 221.27 g/mol
Melting Pt: 103 °C
MDL Number: MFCD00145014
CAS Number: 17639-64-4
https://us.vwr.com/stibo/web/std.lang.all/54/09/28995409.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR