1,6,20,25-Tetraaza[6.1.6.1]paracyclophane ≥98.0% (by HPLC)
Supplier: TCI America
T1378-100MG
TCT1378-100MGEA
208
USD
TCT1378-100MG
1,6,20,25-Tetraaza[6.1.6.1]paracyclophane ≥98.0% (by HPLC)
1,6,20,25-Tetraaza[6.1.6.1]paracyclophane
CAS Number: 74043-83-7
MDL Number: MFCD00142567
Molecular Formula: C34H40N4
Molecular Weight: 504.72
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 183
Formula:
C₃₄H₄₀N₄ Melting Pt: 183 °C |
MDL Number:
MFCD00142567 CAS Number: 74043-83-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...