Tetraphenylphosphonium tetraphenylborate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T1329-25G
TCT1329-025GEA 142.05 USD
TCT1329-025G
Tetraphenylphosphonium tetraphenylborate ≥98.0% (by HPLC, titration analysis)
Tetraphenylphosphonium tetraphenylborate

CAS Number: 15525-15-2
MDL Number: MFCD00145389
Molecular Formula: C48H40BP
Molecular Weight: 658.63
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 306
Flash Point (°C): 211

Formula: C₄₈H₄₀BP
MW: 658.6160 g/mol
Melting Pt: 306 °C
Flash Pt: 211 °C
MDL Number: MFCD00145389
CAS Number: 15525-15-2
https://us.vwr.com/stibo/web/std.lang.all/53/60/28995360.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR