Thiazolidine-2,4-dicarboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Tetrahydrothiazole-2,4-dicarboxylic acid
T1325-5G
TCT1325-005GEA
162.8
USD
TCT1325-005G
Thiazolidine-2,4-dicarboxylic acid ≥98.0% (by HPLC, titration analysis)
Thiazolidine-2,4-dicarboxylic acid
CAS Number: 30097-06-4
MDL Number: MFCD00145399
Molecular Formula: C5H7NO4S
Molecular Weight: 177.17
Purity/Analysis Method:
Form: Crystal
Color: White
Other Product Names / Synonyms: Tetrahydrothiazole-2,4-dicarboxylic Acid
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...