Tetrabutylammonium dichloroaurate
Supplier: TCI America
T1273-100MG
TCT1273-100MGEA
201
USD
TCT1273-100MG
Tetrabutylammonium dichloroaurate
Tetrabutylammonium dichloroaurate
CAS Number: 50480-99-4
MDL Number: MFCD00059125
Molecular Formula: C16H36AuCl2N
Molecular Weight: 510.34
Form: Crystal
Color: White
Melting point (°C): 83
Formula:
C₁₆H₃₆AuCl₂N Melting Pt: 83 °C |
MDL Number:
MFCD00059125 CAS Number: 50480-99-4 |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...