Tetrabutylammonium dibromoaurate
Supplier: TCI America
T1261-1G
TCT1261-001GEA
740
USD
TCT1261-001G
Tetrabutylammonium dibromoaurate
Tetrabutylammonium dibromoaurate
CAS Number: 50481-01-1
MDL Number: MFCD00059119
Molecular Formula: C16H36AuBr2N
Molecular Weight: 599.25
Form: Crystal
Color: White
Formula:
C₁₆H₃₆AuBr₂N |
MDL Number:
MFCD00059119 CAS Number: 50481-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...