1,3,3-Trimethylindolinonaphthospirooxazine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1,3,3-Trimethylspiro[indoline-2,3'-[3H]naphth[2,1-b][1,4]oxazine]
[Photochromic Compound]
CAS Number: 27333-47-7
MDL Number: MFCD00013318
Molecular Formula: C22H20N2O
Molecular Weight: 328.42
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 142
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Photochromic Compounds
Other Product Names / Synonyms: 1,3,3-Trimethylspiro[indoline-2,3'-[3H]naphth[2,1-b][1,4]oxazine]
Formula:
C₂₂H₂₀N₂O MW: 328.42 g/mol Melting Pt: 142 °C |
MDL Number:
MFCD00013318 CAS Number: 27333-47-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...