2,2,5,6-Tetramethyl-4H-1,3-dioxin-4-one ≥95.0%
Supplier: TCI America
T1222-5G
T1222-25G
TCT1222-005GEA
250
USD
TCT1222-005G
TCT1222-25G
2,2,5,6-Tetramethyl-4H-1,3-dioxin-4-one ≥95.0%
2,2,5,6-Tetramethyl-4H-1,3-dioxin-4-one
CAS Number: 87769-39-9
MDL Number: MFCD00060139
Molecular Formula: C8H12O3
Molecular Weight: 156.18
Purity/Analysis Method:
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 72
Specific Gravity (20/20): 1.07
Storage Temperature: 0-10°C
Formula:
C₈H₁₂O₃ Boiling Pt: 72 °C Density: 1.07 (20 °C) |
MDL Number:
MFCD00060139 CAS Number: 87769-39-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...