Trientine tetrahydrochloride 60.0%
Supplier: TCI America
Synonyms:
Triethylenetetramine Tetrahydrochloride
T1212-25G
TCT1212-025GEA
111
USD
TCT1212-025G
Trientine tetrahydrochloride 60.0%
Trientine tetrahydrochloride
CAS Number: 4961-40-4
MDL Number: MFCD00012890
Molecular Formula: C6H18N4
Molecular Weight: 292.07
Purity/Analysis Method:
Form: Crystal
Color: White
Formula:
C₆H₂₂Cl₄N₄ MW: 292.08 g/mol |
MDL Number:
MFCD00012890 CAS Number: 4961-40-4 EINECS: 225-604-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...