Trimesic acid triallyl ester ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1,3,5-Benzenetricarboxylic acid triallyl ester

T1203-5G
TCT1203-005GEA 171.46 USD
TCT1203-005G
Trimesic acid triallyl ester ≥98.0% (by titrimetric analysis)
Trimesic acid triallyl ester

CAS Number: 17832-16-5
MDL Number: MFCD00008647
Molecular Formula: C18H18O6
Molecular Weight: 330.34
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 212
Flash Point (°C): 110
Freezing point (°C): 27


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,3,5-Benzenetricarboxylic Acid Triallyl Ester, Trimesic Acid Triallyl Ester, Triallyl Trimesate

Formula: C₁₈H₁₈O₆
Boiling Pt: 212 °C
Flash Pt: 110 °C
MDL Number: MFCD00008647
CAS Number: 17832-16-5
https://us.vwr.com/stibo/web/std.lang.all/52/51/28995251.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR