Chlorotriphenylgermane ≥97.0%

Supplier: TCI America

Synonyms: ChlorotriphenylgermaneTriphenylgermanium chloride

T1162-1G
TCT1162-001GEA 216.2 USD
TCT1162-001G
Chlorotriphenylgermane ≥97.0%
Chlorotriphenylgermane

CAS Number: 1626-24-0
MDL Number: MFCD00000461
Molecular Formula: C18H15ClGe
Molecular Weight: 339.40
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 118


Other Product Names / Synonyms: Triphenylgermanium Chloride

Formula: C₁₈H₁₅ClGe
Melting Pt: 118 °C
MDL Number: MFCD00000461
CAS Number: 1626-24-0
EINECS: 216-617-5
https://us.vwr.com/stibo/web/std.lang.all/52/18/28995218.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR