2-(3-Thienyl)pyridine ≥95.0%

Supplier: TCI America

Synonyms: 2-(Thiophen-3-yl)pyridine3-(2-Pyridyl)thiophene

T1151-5G
TCT1151-005GEA 803.53 USD
TCT1151-005G
2-(3-Thienyl)pyridine ≥95.0%
2-(3-Thienyl)pyridine

CAS Number: 21298-55-5
MDL Number: MFCD00006214
Molecular Formula: C9H7NS
Molecular Weight: 161.22
Purity/Analysis Method: 95.0% (GC)
Form: Lump
Color: Very Pale Yellow
Freezing point (°C): 25


Other Product Names / Synonyms: 3-(2-Pyridyl)thiophene

Formula: C₉H₇NS
MW: 161.22 g/mol
MDL Number: MFCD00006214
CAS Number: 21298-55-5
https://us.vwr.com/stibo/web/std.lang.all/52/09/28995209.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR