Tetrakis(decyl)ammonium bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tetrakis(decyl)ammonium bromideIPC-TDA-BrTetra(decyl)ammonium bromide

T1142-25G
TCT1142-025GEA 176.11 USD
TCT1142-025G
Tetrakis(decyl)ammonium bromide ≥98.0% (by titrimetric analysis)
Tetrakis(decyl)ammonium bromide

CAS Number: 14937-42-9
MDL Number: MFCD00043166
Molecular Formula: C40H84BrN
Molecular Weight: 659.02
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 88


Other Product Names / Synonyms: Tetrakis(decyl)ammonium Bromide

Formula: C₄₀H₈₄BrN
MW: 659.0057 g/mol
Melting Pt: 88 °C
MDL Number: MFCD00043166
CAS Number: 14937-42-9
EINECS: 239-013-3
https://us.vwr.com/stibo/web/std.lang.all/52/02/28995202.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR