(±)-δ-Tetradecalactone ≥98.0%
Supplier: TCI America
Synonyms:
6-Nonyltetrahydro-2H-pyran-2-one
, (±)-Tetrahydro-6-nonyl-2H-pyran-2-one
, (±)-Tetradecan-5-olide
, δ-Tetradecalactone
CAS Number: 2721-22-4
MDL Number: MFCD00059809
Molecular Formula: C14H26O2
Molecular Weight: 226.36
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 175
Flash Point (°C): 113
Freezing point (°C): 9
Specific Gravity (20/20): 0.94
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: delta-Myristolactone, delta-Tetradecalactone
Formula:
C₁₄H₂₆O₂ MW: 226.36 g/mol Boiling Pt: 175 °C Density: 0.94 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00059809 CAS Number: 2721-22-4 EINECS: 220-334-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...