Tridecanophenone ≥96.0%

Supplier: TCI America

T1087-10G
TCT1087-010GEA 50 USD
TCT1087-010G
Tridecanophenone ≥96.0%
Tridecanophenone

CAS Number: 6005-99-8
MDL Number: MFCD00059219
Molecular Formula: C19H30O
Molecular Weight: 274.45
Purity/Analysis Method: 96.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 42


Other Product Names / Synonyms: Dodecyl Phenyl Ketone

Formula: C₁₉H₃₀O
Melting Pt: 42 °C
MDL Number: MFCD00059219
CAS Number: 6005-99-8
https://us.vwr.com/stibo/web/std.lang.all/51/56/28995156.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR