1,1,3,3-Tetramethylbutyl isocyanide ≥95.0%
Supplier: TCI America
Synonyms:
1,1,3,3-Tetramethylbutylisocyanide
T1054-5ML
T1054-1ML
TCT1054-001MLEA
50
USD
TCT1054-001ML
TCT1054-005ML
1,1,3,3-Tetramethylbutyl isocyanide ≥95.0%
1,1,3,3-Tetramethylbutyl isocyanide
CAS Number: 14542-93-9
MDL Number: MFCD00000003
Molecular Formula: C9H17N
Molecular Weight: 139.24
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 50
Flash Point (°C): 52
Specific Gravity (20/20): 0.80
Storage Temperature: 0-10°C
Formula:
C₉H₁₇N MW: 139.2380 g/mol Boiling Pt: 50 °C Density: 0.80 (20 °C) Flash Pt: 52 °C |
MDL Number:
MFCD00000003 CAS Number: 14542-93-9 UN: 3275 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...