Tetrahydrofurfuryl propionate ≥97.0%

Supplier: TCI America

Synonyms: (Tetrahydrofuran-2-yl)methyl propionate

T1016-25ML T1016-500ML
TCT1016-500MLEA 473 USD
TCT1016-500ML TCT1016-025ML
Tetrahydrofurfuryl propionate ≥97.0%
Tetrahydrofurfuryl propionate

CAS Number: 637-65-0
MDL Number: MFCD00047080
Molecular Formula: C8H14O3
Molecular Weight: 158.20
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 205
Flash Point (°C): 92
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: Propionic Acid Tetrahydrofurfuryl Ester

Formula: C₈H₁₄O₃
MW: 158.1950 g/mol
Boiling Pt: 205 °C
Density: 1.04 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00047080
CAS Number: 637-65-0
https://us.vwr.com/stibo/web/std.lang.all/50/98/28995098.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR