1,1'-Thiobis(2-naphthol) ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Bis(2-hydroxy-1-naphthyl) Sulphide
, 1,1'-Thiodi(2-naphthol)
, Bis(2-hydroxy-1-naphthyl) Sulfide
, 1,1'-Thiobis(naphthalen-2-ol)
CAS Number: 17096-15-0
MDL Number: MFCD00046469
Molecular Formula: C20H14O2S
Molecular Weight: 318.39
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 216
Other Product Names / Synonyms: Bis(2-hydroxy-1-naphthyl) Sulfide, 1,1'-Thiodi(2-naphthol)
Formula:
C₂₀H₁₄O₂S MW: 318.39 g/mol Melting Pt: 216 °C |
MDL Number:
MFCD00046469 CAS Number: 17096-15-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...