TCIAT0913

T0913-500ML T0913-25ML
TCT0913-025MLEA 32.51 USD
TCT0913-025ML TCT0913-500ML
TCIAT0913
CAS Number: 25103-09-7
MDL Number: MFCD00022088
Molecular Formula: C10H20O2S
Molecular Weight: 204.33
Purity/Analysis Method: 90.0% (T)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 128
Specific Gravity (20/20): 0.97

Other Product Names / Synonyms: Thioglycolic Acid Isooctyl Ester, Mercaptoacetic Acid Isooctyl Ester
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR