2-(p-Toluenesulfonyl)acetophenone ≥98.0%
Supplier: TCI America
T0908-10G
TCT0908-010GEA
141.9
USD
TCT0908-010G
2-(p-Toluenesulfonyl)acetophenone ≥98.0%
2-(p-Toluenesulphonyl)acetophenone
CAS Number: 31378-03-7
MDL Number: MFCD00026002
Molecular Formula: C15H14O3S
Molecular Weight: 274.33
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 110
Formula:
C₁₅H₁₄O₃S Melting Pt: 110 °C |
MDL Number:
MFCD00026002 CAS Number: 31378-03-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...